Learn More
Boston Bioproducts Inc Sodium Phosphate Buffer (0.5 M, pH 8.0) - 250ml
Supplier:  Boston Bioproducts Inc BB2328250ML
Encompass_Preferred
The Sodium Phosphate Buffer is a versatile reagent with a high buffering capacity, commonly used across various scientific disciplines across biochemistry and molecular biology. This buffer has a pH range of 5.8 to 8.0. It's applications include protein analysis, where it supports solubility, stability, and activity. It is also compatible with biologically relevant metal ions that are significant for cellular or physiological conditions.
Catalog No. NC1711691
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.