Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate), Reagents

Catalog No. 01337695
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
12 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-695 12 kg
01-337-696 100 g
01-337-694 2.5 kg
01-337-693 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-695 Supplier Reagents Holdings Llc Supplier No. C132405012A5
Add to Cart
Add to Cart

Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Anhydrous, ACS Reagent (Disodium Hydrogen Phosphate)
Color Colorless/White
CAS 7558-79-4
pH 8.4 to 9.6
Physical Form Solid
Quantity 12 kg
Molecular Formula Na2HPO4
Synonym Disodium Hydrogen Phosphate
Molecular Weight (g/mol) 141.9588
Grade ACS
Packaging Poly Pail
Specific Gravity 2.066
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.