Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate), Reagents

Catalog No. 01336985
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-985 500 g
01-336-986 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-985 Supplier Reagents Holdings Llc Supplier No. C2324000500A5
Add to Cart
Add to Cart

Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym Disodium Hydrogen Phosphate
IUPAC Name disodium hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Reagent
Packaging Poly Bottle
Specific Gravity 2.066
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous, Purified (Disodium Hydrogen Phosphate)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.