Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate dibasic dihydrate, meets analytical spec. of BP, Ph. Eur., 98.5-101% (calc. to the dried substance), Solstice

Catalog No. 60044547 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
5 kg
4 x 5 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-547 6 x 1 kg
60-018-36 1 kg
60-018-37 2.5 kg
60-018-38 5 kg
60-044-546 4 x 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-044-547 Supplier Solstice Supplier No. 042726X1KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Meets analytical specification of BP, Ph. Eur., 98.5-101% (calc. to the dried substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10028-24-7
Molecular Formula H5Na2O6P
Molecular Weight (g/mol) 177.99
MDL Number MFCD00149182
InChI Key KDQPSPMLNJTZAL-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate dihydrate, disodium phosphate dihydrate, sodium phosphate dibasic dihydrate, di-sodium hydrogen phosphate dihydrate, phosphoric acid, disodium salt, dihydrate, sodium hydrogenphosphate dihydrate, sodium phosphate, dibasic, dihydrate, disodium hydrogenphosphate dihydrate, dinatrii phosphas dihydricus, 2na.hpo4.2h2o
PubChem CID 6451167
SMILES O.O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 6 x 1 kg
Linear Formula Na2HPO4 · 2H2O
IUPAC Name disodium;hydrogen phosphate;dihydrate
Formula Weight 177.99
Percent Purity 98.5 to 101% (calc. to the dried substance)
Grade Not Available
Chemical Name or Material Sodium Phosphate Dibasic Dihydrate, Meets analytical specification of Ph.Eur.BP
EINECSNumber 231-448-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.