Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), Reagents

Catalog No. 01337698
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-698 2.5 kg
01-337-699 10 kg
01-337-697 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-698 Supplier Reagents Holdings Llc Supplier No. C13240002.5KG
Add to Cart
Add to Cart

Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Dibasic Heptahydrate, ACS Reagent (Disodium Hydrogen Phosphate Heptahydrate)
Color Colorless/White
CAS 7782-85-6
Physical Form Solid
Quantity 2.5 kg
Molecular Formula Na2HPO4, 7H2O
Synonym Disodium Hydrogen Phosphate Heptahydrate
Molecular Weight (g/mol) 268.0658
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.