Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate, Dibasic, Heptahydrate, MP Biomedicals™

Catalog No. MP021914415 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021914415 500 g
MP021914411 100 g
MP219144105 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP021914415 Supplier MP Biomedicals, Inc Supplier No. 02191441.5
Add to Cart
Add to Cart

Widely used in molecular biology, biochemistry and chromatography. Sodium Phosphate, Dibasic, Heptahydrate, MP Biomedicals is a reagent with very high buffering capacity. It is highly hygroscopic, water soluble and useful (with Sodium Phosphate Monobasic)

Sodium phosphate dibasic heptahydrate is use in many applications such as:

  • Sequestrant, emulsifier and buffer in foods
  • As mordant in dyeing, for weighting silk and in tanning
  • In manufacturing of enamels, ceramics, detergents
  • As a fireproofing agent in soldering and brazing instead of borax
  • As a reagent and buffer in analytical chemistry, cathartic, laxative
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-85-6
Molecular Formula H15Na2O11P
Molecular Weight (g/mol) 268.06
MDL Number MFCD00149180
InChI Key LVXHNCUCBXIIPE-UHFFFAOYSA-L
Synonym sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
PubChem CID 6096963
IUPAC Name disodium phosphoric acid heptahydrate
SMILES O.[Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 500 g
Solubility Information Soluble in water, but more soluble in hot water; insoluble in alcohol, ethanol.
Formula Weight 268.07
Percent Purity ≥98%
Grade ACS
Recommended Storage Room Temperature (15–30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.