Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic, MP Biomedicals™

Catalog No. MP219980201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP219980201 1 kg
MP21998021 100 g
MP21998025 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP219980201 Supplier MP Biomedicals, Inc Supplier No. 0219980201
Add to Cart
Add to Cart

Highly hygroscopic and water soluble. Sodium Phosphate Dibasic, MP Biomedicals is a sodium salt of phosphoric acid. This white powder is an important component of running buffer for denaturing gel electrophoresis.

Sodium phosphate dibasic anhydrous

  • Used for preparing phosphate buffer, which in turn is utilized in X-gal staining
  • Induces glomerular calcification as well as other nephrotic syndrome in Sprague-Dawley rats
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Physical Form Crystalline Powder
Quantity 1 kg
Solubility Information Freely soluble in water; insoluble in alcohol
IUPAC Name disodium hydrogen phosphate
Formula Weight 141.98
Percent Purity 98.0 to 100.5%
Grade USP
Recommended Storage Room temperature, desiccate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.