Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium phosphate dibasic, puriss p.a. ACS Reagent, Solstice

Catalog No. 6002843 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
600-28-43 1 kg
600-28-44 250 g
600-28-45 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 600-28-43 Supplier Solstice Supplier No. 716401KG
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
IUPAC Name disodium;hydrogen phosphate
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Quantity 1 kg
Linear Formula Na2HPO4
Formula Weight 141.96g/mol
Percent Purity ≥99% (T)
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium phosphate dibasic
RTECSNumber WC4500000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.