Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade, 98-103%, Spectrum™ Chemical
SDP

Catalog No. 18606710 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-710 12 kg Poly Pail
18-606-706 500 g Poly Bottle
18-606-708 2.5 kg Poly Bottle
18-606-707 6 x 500 g Poly Bottle
18-606-709 4 x 2.5 kg Poly Bottle
18-606-711 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-606-710 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S139612KG
Add to Cart
Add to Cart

S1396, 10049-21-5, NaH2PO4·H2O

Spectrum™ Chemical Sodium Phosphate Monobasic, Monohydrate, Granular, BiotechGrade is a reagent with very high buffering capacity widely used in molecular biology, biochemistry and chromatography. Spectrum offers highly pure reagents suitable for biochemical research and analysis that are absent of inhibitors.

Chemical Identifiers

CAS 10049-21-5
Molecular Formula H5NaO5P
Molecular Weight (g/mol) 139.00
MDL Number MFCD00149208
InChI Key BBMHARZCALWXSL-UHFFFAOYSA-N
IUPAC Name sodium phosphoric acid hydrate
SMILES O.[Na+].OP(O)(O)=O

Specifications

pH 4.1 to 4.5
CAS Max % 1
Quantity 12 kg
Percent Purity 98 to 103%
Grade Biotech
Odor Odorless
Packaging Poly Pail
Appearance White granules
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.