Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Tribasic, Anhydrous, Technical, Spectrum™ Chemical
SDP

Catalog No. 18606719 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-719 2.5 kg Poly Bottle
18-606-718 500 g Poly Bottle
18-606-720 12 kg Poly Pail
18-606-721 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-719 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S14092.5KG
Only null left
Add to Cart
Add to Cart

S1409, 7601-54-9, Na3PO4

Spectrum™ Chemical Sodium Phosphate Tribasic, Anhydrous, Technical or trisodium phosphate, is used as a cleaning agent and is sufficiently alkaline to saponify grease and oils. Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 7601-54-9
Molecular Formula Na3O4P
Molecular Weight (g/mol) 163.94
InChI Key RYFMWSXOAZQYPI-UHFFFAOYSA-K
IUPAC Name trisodium phosphate
SMILES [Na+].[Na+].[Na+].[O-]P([O-])([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Grade Technical
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.