Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Tribasic Dodecahydrate, ≥98.0% (T), Solstice

Catalog No. 60047409 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-409 250 g Poly bottle
60-047-408 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-409 Supplier Solstice Supplier No. 71911250G

Puriss. p.a., ACS Reagent,≥98.0% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10101-89-0
Molecular Formula H24Na3O16P
Molecular Weight (g/mol) 380.119
MDL Number MFCD00149198
InChI Key ASTWEMOBIXQPPV-UHFFFAOYSA-K
Synonym trisodium phosphate dodecahydrate, sodium phosphate dodecahydrate, sodium phosphate tribasic dodecahydrate, phosphoric acid, trisodium salt, dodecahydrate, unii-b70850qphr, ccris 7322, sodium phosphate, tribasic, dodecahydrate, trisodium dodecahydrate phosphate, phosphoric acid, trisodium salt, dodeahydrate, trisodium phosphate tert dodecahydrate
PubChem CID 61473
IUPAC Name trisodium;phosphate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point ∼75°C
Color White
Physical Form Crystalline
Quantity 250 g
Linear Formula Na3PO4 · 12H2O
UN Number NONH for all modes of transport
Solubility Information 70g/L at 20°C
Formula Weight 380.12g/mol
Percent Purity ≥98.0% (T)
Packaging Poly bottle
Density 1.62g/mL at 25°C; Bulk Density: 900 kg/m3
Chemical Name or Material Sodium phosphate tribasic dodecahydrate
Show More Show Less
Hazard Statement P305 + P351 + P338
RTECSNumber 231-509-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.