Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate), Reagents

Catalog No. 01337691
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-691 2.5 kg
01-337-692 10 kg
01-337-690 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-691 Supplier Reagents Holdings Llc Supplier No. C13245002.5A5
Add to Cart
Add to Cart

Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Sodium Phosphate Tribasic Dodecahydrate, ACS Reagent (Trisodium Phosphate Dodecahydrate)
CAS 10101-89-0
Color Colorless/White
Physical Form Solid
Quantity 2.5 kg
Molecular Formula Na3PO4, 12H2O
Synonym Trisodium Phosphate Dodecahydrate
Molecular Weight (g/mol) 380.124
Grade ACS
Packaging Poly Bottle
Specific Gravity 1.6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.