Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Polytungstate, Sigma-Aldrich

Catalog No. 01258990 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-990 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-990 Supplier MilliporeSigma Supelco Supplier No. 71913100G
Add to Cart
Add to Cart

Sodium polytungstate (SPT) is widely used to produce heavy liquid for sink/float analysis (gravity separation) and density gradient centrifugation due to its very high solubility in water.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12141-67-2
Molecular Weight (g/mol) 2986.01 g/mol

Specifications

Percent Purity ≥85%
pH 3
Quantity 100 g
Chemical Name or Material Sodium polytungstate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.