CAS RN 12141-67-2
IUPAC Name:
hexasodium tris(dioxotungstenbis(olate)) nonakis(trioxotungsten)
Synonyms:
hexasodium nonakis(tungsten trioxide) tritungstate
Molecular Weight (g/mol):
2967.98
Molecular Formula:
Na6O39W12
InChi Key:
AZCSOJKJFMWYCX-UHFFFAOYSA-N
SMILES:
[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O.[O-][W]([O-])(=O)=O
1
–
4
of
4
results
1
Sodium metatungstate, 2.82 density solution in water, stabilized
CAS: 12141-67-2 Molecular Formula: H2Na6O40W12 MDL Number: MFCD00150663 Synonym: Sodium polytungstate
3
Sodium Polytungstate, Sigma-Aldrich
CAS: 12141-67-2 Molecular Weight (g/mol): 2986.01 g/mol
4
R&D Systems™ POM 1
CAS: 12141-67-2 Molecular Formula: H2O40Na6W12