Learn More
Invitrogen™ Sodium Potassium ATPase Recombinant Rabbit Monoclonal Antibody (ST0533)
Shop All Thermo Scientific Products

Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | Sodium Potassium ATPase |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | ST0533 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | ATP1A1 |
| Gene Accession No. | P05023, P06685, Q13733, Q64541, Q8VDN2, Q9WV27 |
| Gene Alias | ATP1A1; ATP1A4; ATP1AL2; Atpa-1; ATPase Na+/K+ transporting subunit alpha 1; ATPase Na+/K+ transporting subunit alpha 4; ATPase, Na+/K+ transporting, alpha 1 polypeptide; ATPase, Na+/K+ transporting, alpha 4 polypeptide; ATPase, Na+/K+ transporting, alpha polypeptide-like 2; ATPase, Na+K+ transporting, alpha 1; BC010319; Na(+)/K(+) ATPase alpha-1 subunit; Na(+)/K(+) ATPase alpha-4 subunit; Na, K-ATPase, alpha-A catalytic polypeptide; Na,K-ATPase alpha-1 subunit; Na,K-ATPase catalytic subunit alpha-A protein; Na,K-ATPase subunit alpha-C; Na+/K+ ATPase 1; Na+/K+ ATPase 4; Na+/K+ ATPase alpha 4 subunit; Na+/K+ ATPase, alpha-D polypeptide; Nkaa1b; sodium pump 4; sodium pump subunit alpha-1; Sodium pump subunit alpha-4; sodium/potassium-transporting ATPase alpha-4 chain; sodium/potassium-transporting ATPase subunit alpha-1; Sodium/potassium-transporting ATPase subunit alpha-4; sodium-potassium ATPase catalytic subunit alpha-1; sodium-potassium ATPase catalytic subunit alpha-4; sodium-potassium-ATPase, alpha 1 polypeptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.