Learn More
Sodium Potassium Tartrate, For Nitrogen (Ammonia), Nitrogen (Organic), Certified, 10% (w/v), LabChem™
Shop All LabChem ProductsSpecifications
Specifications
| Color | Colorless |
| CAS Max % | 89.7, 10, 0.2, 0.05, 0.05 |
| Linear Formula | NaKC4H4O6·4H2O |
| Solubility Information | Soluble in water |
| Formula Weight | 282.23 |
| Percent Purity | 10% (w/v) |
| Grade | Certified |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Decomposition Information | Carbon monoxide; Carbon dioxide |
| Show More |
Safety and Handling
| Hazard Statement | GHS H Statement Solution is not hazardous. |
| Precautionary Statement | GHS P Statement If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes. If swallowed, get medical attention. |
| Recommended Storage | Refrigerate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.