Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pyridine-3-sulfonate 97.0+%, TCI America™
SDP

Catalog No. P061825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P061825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P061825G Supplier TCI America Supplier No. P061825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15521-77-4
Molecular Formula C5H4NNaO3S
Molecular Weight (g/mol) 181.14
MDL Number MFCD00065041
InChI Key CQSNULMCHIOCCX-UHFFFAOYSA-M
Synonym Pyridine-3-sulfonic Acid Sodium Salt
PubChem CID 23677935
IUPAC Name sodium pyridine-3-sulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CN=CC=C1

Specifications

Color White
Quantity 25 g
Formula Weight 181.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pyridine-3-sulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.