Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium pyrophosphate decahydrate, ACS, 99.0-103.0%

Catalog No. AA3338536
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3338536 500 g
AA3338522 100 g
AA33385A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3338536 Supplier Thermo Scientific Chemicals Supplier No. 03338536
Add to Cart

CAS: 13472-36-1 | H20Na4O17P2 | 446.05 g/mol

Sodium pyrophosphate decahydrate is used as a buffering agent, an emulsifier, a thickening agent, a food additive and a dispersing agent. It acts as a tartar control agent in tooth paste and dental floss and is used to remove calcium and magnesium from saliva, thereby avoiding its deposition on teeth. It is utilized in detergents to prevent deposition on cloth. It is useful in the preparation of an EDTA-sodium pyrophosphate extraction buffer for microcystin analysis of soil samples. In antimicrobial studies, it is used as a chelating agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13472-36-1
Molecular Formula H20Na4O17P2
Molecular Weight (g/mol) 446.05
MDL Number MFCD00149200
InChI Key VZWGHDYJGOMEKT-UHFFFAOYSA-J
Synonym sodium pyrophosphate decahydrate, unii-iy3dkb96qw, tetrasodium pyrophosphate decahydrate, diphosphoric acid, tetrasodium salt, decahydrate, iy3dkb96qw, pyrophosphate sodium salt decahydrate, pyrophosphate inorganic, colostrum, sodium phosphate pyro, sodium pyrophosphate na4p2o7 decahydrate
PubChem CID 3084150
IUPAC Name tetrasodium;phosphonato phosphate;decahydrate
SMILES O.O.O.O.O.O.O.O.O.O.[O-]P(=O)([O-])OP(=O)([O-])[O-].[Na+].[Na+].[Na+].[Na+]

Specifications

Melting Point 79.5°C
Color White
Physical Form Crystalline
Quantity 500 g
Assay Percent Range 99.0 to 103.0%
Solubility Information Soluble in water. Insoluble in alcohol and ammonia.
Formula Weight 446.06 (265.90 Anhydrous)
Grade ACS
Density 1.82 g/mL
Chemical Name or Material Sodium pyrophosphate decahydrate, For analysis ACS
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-767-1
RTECSNumber UX7350000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.