Learn More
Apexbio Technology LLC Oxacillin sodium monohydrate 7240-38-2 10mM (in 1mL H2O)
SDP
Supplier:  Apexbio Technology LLC B19975.1
Encompass_Preferred
Oxacillin sodium monohydrate (CAS 7240-38-2) is a small-molecule inhibitor within the penicillin class It is designed to inhibit bacterial cell wall biosynthesis thereby disrupting peptidoglycan cross-linking and compromising cellular integrity Oxacillin sodium monohydrate exerts its biological activity primarily through interference with peptidoglycan cross-linking resulting in bacterial lysis In in vitro studies Oxacillin sodium monohydrate demonstrates antimicrobial inhibition with minimum inhibitory concentration (IC50) values typically ranging from approximately 0 1 to several g/mL depending on bacterial strain and assay conditions Based on these pharmacological properties Oxacillin sodium monohydrate holds research potential in antimicrobial susceptibility testing resistance phenotyping and investigations of bacterial resistance mechanisms particularly in gram-positive -lactamase-producing bacteria such as methicillin-resistant staphylococci (MRSA)
Catalog No. 50-000-93220
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.