Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Sodium Salt, Pipes, OmniPur™, Calbiochem™,

Catalog No. 69121KG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-121-KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-121-KG Supplier MilliporeSigma™ Supplier No. 69121KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular biology grade PIPES sodium salt with ≥99.0% purity.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100037-69-2
Molecular Formula C16H33N4Na3O12S4
Molecular Weight (g/mol) 670.665
InChI Key FPDACHMSOMILQI-UHFFFAOYSA-K
PubChem CID 3086462
IUPAC Name trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

For Use With (Application) Molecular Biology
Physical Form Fine Crystalline Powder
Quantity 1 kg
DNase None Detected
Assay Percent Range ≥99.0%
Protease None Detected
Formula Weight 335.37
Percent Purity ≥99.0%
Grade OmniPur
Chemical Name or Material Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.