Learn More
MilliporeSigma™ PIPES, Sodium Salt, Molecular biology grade, OmniPur™, Calbiochem™,
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Color | White |
| For Use With (Application) | Molecular Biology |
| Physical Form | Fine Crystalline Powder |
| Quantity | 250 g |
| Assay Percent Range | ≥99% |
| Percent Purity | ≥99.0% (anhydrous basis) |
| Grade | Molecular Biology Reagent |
| Chemical Name or Material | Sodium Salt |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.