Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium selenate, Purum p.a., ≥98.0% (T), Solstice

Catalog No. 60047415 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-047-415 100 g
60-015-50 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-415 Supplier Solstice Supplier No. 71948100G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Purum p.a.,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13410-01-0
Molecular Formula Na2O4Se
Molecular Weight (g/mol) 188.95
MDL Number MFCD00003490
InChI Key MHQOTKLEMKRJIR-UHFFFAOYSA-L
Synonym sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
PubChem CID 25960
ChEBI CHEBI:77775
IUPAC Name disodium;selenate
SMILES [Na+].[Na+].[O-][Se]([O-])(=O)=O

Specifications

CAS Max % ≤100.0000%
Quantity 100 g
Linear Formula Na2SeO4
UN Number UN2630
Formula Weight 188.94
Percent Purity ≥98%
Grade purum p.a.
Packaging Poly Bottle
Density 3.213 g/cm3
Chemical Name or Material Sodium selenate, Purum p.a., ≥98.0% (T)
Hazard Statement P261-P273-P301 + P310-P311-P501
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 6.1; Packing Group:3: I; Proper Shipping Name: Selenates
EINECSNumber 236-501-8
RTECSNumber 236-501-8
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.