Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenite, MP Biomedicals™

Catalog No. 0219474190 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-74190 500 g
ICN19474110 10 g
ICN19474180 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-194-74190 Supplier MP Biomedicals, Inc Supplier No. 0219474190
Add to Cart
Add to Cart

Sodium Selenite is a source of selenium in biological research, where selenium is an essential trace element that is normally provided by serum.

  • Selenium is present in selenoproteins such as glutathione peroxidase and thioredoxin reductase, which contain the selenium analog of cysteine, selenocysteine.
  • In particular, glutathione peroxidase has a role in detoxification in vivo as a scavenger of peroxides.
  • Sodium selenite is included in various medium supplements for use in cell culture.
  • Sodium selenite has been utilized in studies of cell proliferation and cancer research.
  • It has been used to alter gene expression in HepG2 cells as analyzed by cDNA microarrays.
  • Sodium selenite can inhibit zinc finger protein/DNA interactions.
  • Sodium selenite has been shown to alter mitochondrial membrane potentials and thus potentially contribute to apoptosis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10102-18-8
Molecular Formula Na2O3Se
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L

Specifications

Boiling Point Decomposition
Melting Point 1310°F (Decomposition)
Quantity 500 g
SMILES [Na+].[Na+].[O-][Se]([O-])=O
IUPAC Name disodium selenite
Molecular Weight (g/mol) 172.95
Percent Purity ∼98%
Grade Cell Culture
Chemical Name or Material Sodium Selenite
Recommended Storage 15 to 30°C

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.