Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Selenite Pentahydrate, Solstice

Catalog No. 6001551 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-51 100 g
60-015-52 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-015-51 Supplier Solstice Supplier No. 00163100G
Add to Cart
Add to Cart

Puriss. p.a.,≥99.0% (RT)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26970-82-1
Molecular Formula H10Na2O8Se
Molecular Weight (g/mol) 263.023
MDL Number MFCD00149173
InChI Key TUANAMBRHOLYTH-UHFFFAOYSA-L
Synonym sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
PubChem CID 134930
IUPAC Name disodium;selenite;pentahydrate
SMILES O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]

Specifications

Quantity 100 g
Linear Formula Na2SeO3 · 5H2O
UN Number 2630
Formula Weight 263.01g/mol
Percent Purity ≥99% (RT)
Chemical Name or Material Sodium Selenite Pentahydrate
Hazard Statement P261-P273-P280-P301 + P310 + P330-P304 + P340 + P312-P403 + P233
Precautionary Statement H300-H317-H331-H411
EINECSNumber 233-267-9
RTECSNumber VS7420000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.