Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Silicofluoride, 95%, Spectrum™ Chemical
SDP

Catalog No. 18613858 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-858 100 g Poly Bottle
18-613-860 500 g Poly Bottle
18-613-861 100 lb. Fiber Drum
18-613-862 2.5 kg Poly Bottle
18-613-859 12 kg Poly Pail
18-613-857 25 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-613-858 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1440100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16893-85-9

Specifications

CAS Max % 1
Quantity 100 g
Percent Purity 95%
Grade Reagent
Loss on Drying 0.005
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.