Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sorbate 98.0+%, TCI America™
SDP

Catalog No. S0058500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0058500G 500 g
S005825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0058500G Supplier TCI America Supplier No. S0058500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7757-81-5
Molecular Formula C6H7NaO2
Molecular Weight (g/mol) 134.11
MDL Number MFCD00058995
InChI Key LROWVYNUWKVTCU-STWYSWDKSA-M
Synonym Sorbic Acid Sodium Salt
PubChem CID 23665582
IUPAC Name sodium;(2E,4E)-hexa-2,4-dienoate
SMILES CC=CC=CC(=O)[O-].[Na+]

Specifications

Color White-Yellow
Physical Form Crystalline Powder
Quantity 500 g
Formula Weight 134.11
Percent Purity ≥98.0% (T)
Chemical Name or Material Sodium Sorbate
EINECSNumber (2)-0611
RTECSNumber WG2220000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.