Learn More
Sodium Standard, Inorganic, 10,000 μg/mL, 125 mL
Specifications
Specifications
| CAS Max % | 97.0, 2.0, 1.0 |
| Analyte or Component Names | Sodium,water,nitric acid |
| Element | Nitric Acid, Na |
| Solvent or Matrix | Water/Dilute Nitric Acid |
| Quantity | 125 mL |
| Concentration or Composition (by Analyte or Components) | 10, 000μg/mL |
| Chemical Name or Material | Sodium Standard |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.