Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Stearate, Powder, NF, 90%, Spectrum™ Chemical
SDP

Catalog No. 18606797 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-797 500 g Poly Pail
18-606-798 2.5 kg Poly Pail
18-606-800 45 kg Fiber Drum
18-606-799 12 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-797 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1447500GM
Add to Cart
Add to Cart

S1447, 822-16-2, C18H35NaO2

Spectrum™ Chemical Sodium Stearate, Powder, NF is used in the pharmaceutical industry as a surfactant to aid the solubility of hydrophobic compounds. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 822-16-2
Molecular Formula C18H35NaO2
Molecular Weight (g/mol) 306.47
InChI Key RYYKJJJTJZKILX-UHFFFAOYSA-M
IUPAC Name sodium octadecanoate
SMILES [Na+].CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 90%
Grade NF
Loss on Drying 0.05
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.