Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfanilate dihydrate, 99%, Thermo Scientific™

Catalog No. AC419642500
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC419642500 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC419642500 Supplier Thermo Scientific Chemicals Supplier No. 419642500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-22-5
Molecular Formula C6H10NNaO5S
Molecular Weight (g/mol) 231.198
InChI Key MJFZIDUBUNIFLZ-UHFFFAOYSA-M
Synonym sodium sulfanilate dihydrate, unii-y31oen1192, 4-aminobenzenesulfonic acid, sodium salt dihydrate, sodium 4-aminobenzenesulfonate dihydrate, benzenesulfonic acid,4-amino-, sodium salt, hydrate 1:1:?, sulfanilic acid sodium salt dihydrate mi, acmc-1b9i8, c6h6nso3.na.2h2o, benzenesulfonic acid, 4-amino-, sodium salt, hydrate 1:1:2, 4h-1,4-benzoxazin-4-amine,2,3-dihydro hcl
PubChem CID 23687713
IUPAC Name sodium;4-aminobenzenesulfonate;dihydrate
SMILES C1=CC(=CC=C1N)S(=O)(=O)[O-].O.O.[Na+]

Specifications

Assay Percent Range 99%
Linear Formula SO3NaC6H4NH2·2H2O
Quantity 250 g
Merck Index 15, 9058
Formula Weight 213.19
Chemical Name or Material Sodium sulfanilate dihydrate, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.