Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, 1 M, Filling Solution, Fisherbrand
SDP

Catalog No. ACCU0833500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ACCU0833500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ACCU0833500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 7757-82-6
Molecular Weight (g/mol) 142.04 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Physical Form Liquid
Quantity 500 mL
Grade Laboratory
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Sulfate, 1M, Fill Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.