Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate ACS AR Granular, Macron Fine Chemicals™

Catalog No. MK8024500 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
200 lb.
Packaging:
Fiber Drum
Glass
STAKMOR
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-8024-500 500 g Glass
MK802424 200 lb. Fiber Drum
MK-8024-212 2.5 kg Glass
MK-8024-12 12 kg STAKMOR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. MK-8024-500 Supplier Avantor Supplier No. 802404
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

pH 5.2 to 9.2 (5% solution at 25°C)
Physical Form Granules
Quantity 500 g
Assay Percent Range min. 99.0%
Formula Weight 142.04
Grade For ACS analysis
Loss on Ignition max. 0.5%
Packaging Glass
Appearance White crystalline granules
Chemical Name or Material Sodium Sulfate, Anhydrous, 10-60 Mesh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.