Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous, 10-60 Mesh, MilliporeSigma™

Catalog No. MSX0760E1 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
45 kg
Packaging:
Fiber Drum w/bag
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MSX0760E1 500 g Plastic Bottle
MSX0760E3 2.5 kg Plastic Bottle
MSX0760E20 45 kg Fiber Drum w/bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MSX0760E1 Supplier MilliporeSigma Supplier No. SX0760E1
This item is not returnable. View return policy

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Physical Form Granules
Quantity 500 g
Linear Formula Na2SO4
Formula Weight 142.04
Packaging Plastic Bottle
Chemical Name or Material Sodium Sulfate, Anhydrous, 10-60 Mesh
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.