Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfate anhydrous, For ACS analysis, 99.32%, MP Biomedicals™

Catalog No. ICN19144405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19144405 5 kg
ICN19144401 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19144405 Supplier MP Biomedicals, Inc Supplier No. 0219144405
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Powder
Quantity 5 kg
Assay Percent Range 0.9932
Formula Weight 142.04
Density 2.7g/cm3 at 20°C
Chemical Name or Material Sodium sulfate anhydrous, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.