Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate Anhydrous, MACCO

Catalog No. 502985635
Encompass
Change view
Click to view available options
Quantity:
12 kg
50 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-298-5635 50 kg
50-298-5599 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-298-5635 Supplier Thermo Scientific Production Chemicals and Services Supplier No. M51104000500
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7757-82-6
Molecular Weight (g/mol) 142.04 g/mol

Specifications

Physical Form Crystalline Powder
Quantity 50 kg
Percent Purity 99 to 100%
Grade USP
Packaging 50 kg poly drum with 2xPE liner
Chemical Name or Material Sodium sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.