Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfate, Anhydrous, USP, EP, BP, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606833 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-833 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-833 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO20412KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SO204, 7757-82-6, Na2SO4

Spectrum™ Chemical Sodium Sulfate, Anhydrous, USP, EP, BP is an electrolyte replenisher and is used in isosmotic solutions. All Spectrum Chemical USP, EP, BP productsare manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Physical Form Crystalline Powder, Needles or Crystals
Quantity 12 kg
Percent Purity 99 to 101%
Grade BP/EP/USP
Loss on Drying 0.005
Packaging Poly Pail
Chemical Name or Material Sodium Sulfate, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.