Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, Fused Flakes, Approx. 60%, Spectrum™ Chemical
SDP

Catalog No. 18606846 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
400 lb.
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-846 400 lb. Amber Glass Bottle
18-606-842 500 g Amber Glass Bottle
18-606-843 2.5 kg Amber Glass Bottle
18-606-844 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-606-846 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1470400LBBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1470, 27610-45-3, Na2S·xH2O

Spectrum™ Chemical Sodium Sulfide, Fused Flakes , also known as disodium sulfide, is used as an oxygen scavenger agent in water treatment as well as a metals precipitant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
IUPAC Name disodium sulfanediide
SMILES [Na+].[Na+].[S--]

Specifications

CAS Max % 1
Physical Form Flakes or Powder
Quantity 400 lb.
Percent Purity ∼60%
Grade Ungraded
Packaging Amber Glass Bottle
Chemical Name or Material Sodium Sulfide, Fused Flakes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.