accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaS

CAS RN 27610-45-3

IUPAC Name: disodium sulfanediide
Synonyms: sodium sulfide
Molecular Weight (g/mol): 78.04
Molecular Formula: Na2S
InChi Key: GRVFOGOEDUUMBP-UHFFFAOYSA-N
SMILES: [Na+].[Na+].[S--]
Molecular Weight (g/mol) 
  • (12)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
Grade 
  • (3)
  • (4)
  • (5)
Physical Form 
  • (2)
  • (3)
  • (5)
  • (2)
Percent Purity 
  • (5)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (2)
  • (2)
  • (5)
  • (5)
  • (12)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (5)
  • (2)
  • (3)
  • (5)
  • (2)
  • (5)
  • (3)