Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, Fused Flakes, Technical, Reagents

Catalog No. 01336958
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-958 2.5 kg
01-336-957 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-958 Supplier Reagents Holdings Llc Supplier No. C23260002.5A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Sulfide, Fused Flakes, Technical, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 27610-45-3
Molecular Formula Na2S
Molecular Weight (g/mol) 78.04
MDL Number MFCD00149183
InChI Key GRVFOGOEDUUMBP-UHFFFAOYSA-N
Synonym Sodium sulphide scales
IUPAC Name disodium sulfanediide
SMILES [Na+].[Na+].[S--]

Specifications

Melting Point 50°C
Color Colorless to Slight Yellow
Physical Form Solid
Quantity 2.5 kg
Grade Technical
Packaging Poly Bottle
Specific Gravity 1.856
Chemical Name or Material Sodium Sulfide, Fused Flakes, Technical
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.