Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium sulfide nonahydrate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog No. AA3662230
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3662230 250 g
AA36622A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3662230 Supplier Thermo Scientific Chemicals Supplier No. 03662230

It is the precursor material for preparation of cadmium sulfide quantum dots. Also employed in the pulp and paper industry. It is likewise employed in the making of colors and dyestuffs. It is too useful in the trace quantities for the rapid synthesis of small silver nanocubes through mediating polyol reduction. It has as a significant role in breaking up the feather keratin useful for the preparation of bio-polymer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Melting Point ∽50°C (decomposition)
Physical Form Crystalline
Quantity 250 g
Assay Percent Range 98% min.
Solubility Information Soluble in water; slightly soluble in alcohol. Insoluble in ether.
Formula Weight 240.18 (78.04 Anhydrous)
Grade ACS
Sensitivity Hygroscopic; Light sensitive
Appearance Crystalline
Density 1.427 g/cm3 at 20°C
Chemical Name or Material Sodium sulfide nonahydrate, For analysis ACS
Show More Show Less
missing translation for 'healthHazard1' H290-H302-H311-H314
missing translation for 'healthHazard3' P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 215-211-5
RTECSNumber WE1925000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.