CAS RN 1313-84-4
Sodium Sulfide, Nonahydrate, Nonahydrate, 98.0% min., ACS, MilliporeSigma™
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium Sulfide, 5% (w/v), Ricca Chemical
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium Sulfide, 3% (w/v), Ricca Chemical
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium Sulfide, Solution No. 1 for Zinc, Certified, 3% (w/v), LabChem™
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium Sulfide Nonahydrate, Solstice
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Fisher Science Education™ Sodium Sulfide Nonahydrate
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N Synonym: Sodium Monosulfide Nonahydrate IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sigma Aldrich Sodium sulfide nonahydrate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sodium Sulfide Nonahydrate ACS MP Biomedicals
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N Synonym: Sodium Sulphide,Sodium Sulfide enneahydrate,Disodium Sulfide Nonahydrate IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium sulfide nonahydrate, ACS, 98.0% min, Thermo Scientific Chemicals
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sodium Sulfide, Nonahydrate, Crystal, Reagent, ACS, 98%, Spectrum™ Chemical
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Sulfide in Soil - PT, MilliporeSigma™ Supelco™
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
Sodium Sulfide Nonahydrate p.a., Thermo Scientific™
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]
Fisher Science Education™ Sodium Sulfide Solution, 0.1M
CAS: 1313-84-4 Molecular Formula: H18Na2O9S Molecular Weight (g/mol): 240.17 MDL Number: MFCD00149184 InChI Key: ZGHLCBJZQLNUAZ-UHFFFAOYSA-N IUPAC Name: disodium nonahydrate sulfanediide SMILES: O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]