Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide, Nonahydrate, Crystal, Reagent, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18606852 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 kg
4 x 2.5 kg
Packaging:
Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-852 4 x 2.5 kg Poly Pail
18-606-851 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-606-852 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S14652.5KGCS4

missing translation for 'validMainframeMsg'

S1465, 1313-84-4, Na2S·9H2O

Spectrum™ Chemical Sodium Sulfide, Nonahydrate, Crystal, Reagent, ACS is a colorless chemical compound that is soluble in water. It often appears yellow in color due to the presence of polysulfides, although it is colorless as a solution. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

CAS Max % 1
Quantity 4 x 2.5 kg
Percent Purity 98%
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.