Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Sulfide Nonahydrate, Solstice

Catalog No. 6001557 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-57 500 g
60-015-54 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-015-57 Supplier Solstice Supplier No. 208043500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-84-4
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

Color Colorless or Yellow
Quantity 500 g
Linear Formula Na2S·9H2O
UN Number 1849
Formula Weight 240.18g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Sodium Sulfide Nonahydrate
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
Precautionary Statement H302-H311-H314-H400
EINECSNumber 215-211-5
RTECSNumber WE1925000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.