Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Sulfide Solution, 0.1M
SCI_ED

Catalog No. S25891 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S25891 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25891

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 1313-84-4 , 7732-18-5
Molecular Formula H18Na2O9S
Molecular Weight (g/mol) 240.17
MDL Number MFCD00149184
InChI Key ZGHLCBJZQLNUAZ-UHFFFAOYSA-N
IUPAC Name disodium nonahydrate sulfanediide
SMILES O.O.O.O.O.O.O.O.O.[Na+].[Na+].[S--]

Specifications

CAS Min % 0.0
CAS Max % 2.4, 97.6
Density 1.000
Packaging 0.1M
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Sodium Sulfide Solution, 0.1M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.