Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Tartrate Dibasic Dihydrate

Catalog No. 71995250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-995-250G 250 g
71-995-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-995-250G Supplier Sigma Aldrich Supplier No. 71995250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium tartrate dibasic dihydrate (L-(+)-Tartaric acid disodium salt, Sodium-L-tartrate dibasic dihydrate) is the sodium salt of tartaric acid. Mixtures of dimethyl sulfoxide and 1% aqueous solution of sodium-L-tartrate dibasic dihydrate have been employed for the resolution of various quaternary ammonium surfactants.Sodium tartrate dibasic dihydrate (L-(+)-Tartaric acid disodium salt, Sodium-L-tartrate dibasic dihydrate) may be used for the growth of crystals of xylanase C (XynC) fro mgram-positive bacterium Bacillus subtilis. It may be used to compose the reservoir solution for the growth of crystals of PcpR (a LysR-type transcription factor from Sphingobium chlorophenolicu mL-1).

Specifications

CAS 6106-24-7
Molecular Formula C4H4Na2O6 -+ 2H2O
Linear Formula C4H4Na2O6 -+ 2H2O
MDL Number MFCD00150035
Quantity 250 g
Synonym L-(+)-Tartaric acid disodium salt; Disodium tartrate dihydrate; Sodium tartrate dihydrate
Molecular Weight (g/mol) 230.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.