Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate, Dihydrate, 99.0 to 101.0%, ACS, MilliporeSigma™

Catalog No. MSX07951 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MSX07951 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MSX07951 Supplier MilliporeSigma Supplier No. SX07951
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
PubChem CID 131855972
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Color White or Colorless
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 99.0 to 101.0%
Formula Weight 23.8
Density 1.82g/cm3; Bulk Density: 460 kg/m3
Chemical Name or Material Sodium Tartrate, Dihydrate, EMSURE™, For Analysis
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.