Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate Dihydrate, ACS Reagent, Reagents

Catalog No. 01337667
Change view
Click to view available options
Quantity:
125 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-667 125 g
01-337-664 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-667 Supplier Reagents Holdings Llc Supplier No. C1327000125D2
Add to Cart
Add to Cart

Sodium Tartrate Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

Melting Point 150°C
Color Colorless/White
pH 7 to 9
Physical Form Solid
Quantity 125 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 1.82
Chemical Name or Material Sodium Tartrate Dihydrate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.