Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate Dihydrate (Colorless Crystals), Fisher BioReagents

Catalog No. BP352500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP352-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP352-500
Add to Cart

CAS: 6106-24-7 | C4H8Na2O8 | 230.08 g/mol

Glass Bottle; 500g

This sodium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Tartrate Dihydrate
CAS 6106-24-7, 868-18-8
Color White
pH 7 to 9
Loss on Drying 15.61 to 15.71% (at 150°C)
Packaging Glass Bottle
Molecular Formula C4H8Na2O8
MDL Number MFCD00150035
Quantity 500 g
Merck Index 15, 8812
Synonym unii-dia7c37aow, disodium tartrate dihydrate, disodium l-+-tartrate dihydrate, dia7c37aow, disodium +-tartrate dihydrate, sodium l-+-tartrate dihydrate, butanedioic acid, 2,3-dihydroxy-2r,3r-, disodium salt, dihydrate, disodium tartarate dihydrate, sodium tartrate nf, sodium tartrate dihydrate mi
Solubility Information ≤0.005% Insoluble Matter
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O
IUPAC Name (2R,3R)-2,3-dihydroxybutanedioic acid;sodium;dihydrate
Molecular Weight (g/mol) 230.08
PubChem CID 131855972
Formula Weight 230.08
Percent Purity 99.0 to 101.0%
Physical Form Powder or Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.