Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS, 99-101%, Spectrum™ Chemical
SDP

Catalog No. 18606874 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
25 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-874 45 kg Fiber Drum
18-606-868 500 g Poly Bottle
18-606-870 2.5 kg Poly Bottle
18-606-869 6 x 500 g Poly Bottle
18-606-871 4 x 2.5 kg Poly Bottle
18-606-872 12 kg Poly Pail
18-606-873 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-874 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S148545KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S1485, 6106-24-7, Na2C4H4O6·2H2O

Spectrum™ Chemical Sodium Tartrate, Dihydrate, Crystal, Reagent, ACS has a crystal structure that is able to capture a very precise amount of water and is used in the Karl Fischer moisture determination titration technique. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 6106-24-7
Molecular Formula C4H8Na2O8
Molecular Weight (g/mol) 230.08
MDL Number MFCD00150035
InChI Key FGJLAJMGHXGFDE-UDMGOBQRNA-L
IUPAC Name disodium (2R,3R)-2,3-dihydroxybutanedioate dihydrate
SMILES O.O.[Na+].[Na+].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

Specifications

CAS Max % 1
pH 7.0 to 9.0
Loss on Drying 15.61-15.71%
Packaging Fiber Drum
Quantity 45 kg
Percent Purity 99.0 to 101%
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.