Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium tert-pentoxide, 95%, pure

Catalog No. AC327551000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327551000 100 g Glass bottle
AC327555000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC327551000 Supplier Thermo Scientific Chemicals Supplier No. 327551000
Add to Cart

CAS: 14593-46-5 | C5H11NaO | 110.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14593-46-5
Molecular Formula C5H11NaO
Molecular Weight (g/mol) 110.13
MDL Number MFCD00064245
InChI Key CGRKYEALWSRNJS-UHFFFAOYSA-N
Synonym sodium tert-pentoxide, sodium 2-methylbutan-2-olate, sodium tert-amylate, sodium t-amylate, sodium-t-amylate, unii-51z39py5z8, sodium-tert-pentoxide, sodium tert-amoxide, 2-butanol, 2-methyl-, sodium salt, tert-amyl alcohol sodium salt
PubChem CID 2733478
IUPAC Name sodium;2-methylbutan-2-olate
SMILES CCC(C)(C)[O-].[Na+]

Specifications

Color Cream or Tan
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3CH2C(CH3)2ONa
Quantity 100 g
Formula Weight 110.13
Percent Purity 95%
Grade Pure
Physical Form Powder
Chemical Name or Material Sodium tert-pentoxide, 95%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable solid.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep cool.
Protect from sunlight.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do N
EINECSNumber 238-639-4
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.