Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Tetraphenylborate, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18613892 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-892 25 g Amber Glass Bottle
18-613-891 10 g Amber Glass Bottle
18-613-893 100 g Amber Glass Bottle
18-613-894 1 kg Amber Glass Bottle
18-613-895 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-613-892 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S149025GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-66-8

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99.5%
Grade ACS
Loss on Drying 0.005
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.